Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022689.2 | NP_073180.1 | 718 | 98 | ||
GenBank mRNA | AF052596.1 | - | 570 | 98 | ||
BC127494.1 | - | 718 | 8361815 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |