Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033870.1 | NP_001029042.1 | 11 - 12 | 1652 | 74 | |
NM_023102.2 | NP_075590.2 | 11 - 12 | 1649 | 74 | ||
XM_006241000.2 | XP_006241062.1 | 10 - 11 | 3027 | 74 | ||
XM_006241001.2 | XP_006241063.1 | 10 - 11 | 1735 | 74 | ||
XM_006241002.2 | XP_006241064.1 | 11 - 12 | 2947 | 74 | ||
XM_006241003.2 | XP_006241065.1 | 10 - 11 | 2377 | 74 | ||
XM_008765130.1 | XP_008763352.1 | 10 - 11 | 1735 | 74 | ||
GenBank mRNA | BC072533.1 | - | 11 - 12 | 1652 | 7098992 | 74 |
BC099758.1 | - | 5 - 6 | 511 | 74 | ||
U22297.1 | - | 10 - 11 | 1383 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |