Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023965.1 | NP_076455.1 | 5 - 6 | 489 | 118 | |
GenBank mRNA | AF298656.3 | - | 5 - 6 | 489 | 118 | |
AJ295950.1 | - | 5 - 6 | 489 | 118 | ||
FQ217794.1 | - | 558 | 118 | |||
FQ231233.1 | - | 542 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |