Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023991.1 | NP_076481.1 | 3 - 4 | 338 | 72 | |
XM_008763901.1 | XP_008762123.1 | 2 - 3 | 2067 | 72 | ||
GenBank mRNA | U12149.1 | - | 3 - 4 | 523 | 72 | |
Z29486.1 | - | 3 - 4 | 338 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |