Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031045.2 | NP_112307.2 | 2 - 3 | 625 | 77 | |
GenBank mRNA | M29787.1 | - | 2 - 3 | 625 | 77 | |
X56917.1 | - | 2 - 3 | 616 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |