Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031055.1 | NP_112317.1 | 6 - 7 | 1006 | 117 | |
GenBank mRNA | U24441.1 | - | 6 - 7 | 1006 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |