Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031066.1 | NP_112328.1 | 6 - 7 | 1090 | 82 | |
GenBank mRNA | BC087740.1 | - | 6 - 7 | 1090 | 7314065 | 82 |
FQ212529.1 | - | 4 - 5 | 836 | 82 | ||
U48249.2 | - | 6 - 7 | 1053 | 82 | ||
U63740.1 | - | 6 - 7 | 1090 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |