Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031546.1 | NP_113734.1 | 1 - 2 | 124 | ||
GenBank mRNA | AB037934.1 | - | 1 - 2 | 124 | ||
BC078794.1 | - | 1 - 2 | 78 | 7126233 | 124 | |
D38467.1 | - | 1 - 2 | 124 | |||
FQ219778.1 | - | 1 - 2 | 86 | 124 | ||
X69021.1 | - | 1 - 2 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |