Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053537.2 | NP_445989.2 | 7 - 8 | 1276 | 70 | |
GenBank mRNA | AY816233.1 | - | 7 - 8 | 1276 | 70 | |
L27651.1 | - | 7 - 8 | 1336 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |