Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053595.2 | NP_446047.1 | 5 - 6 | 719 | 68 | |
GenBank mRNA | BC087006.1 | - | 5 - 6 | 719 | 7193034 | 68 |
L40030.1 | - | 5 - 6 | 454 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |