Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053738.1 | NP_446190.1 | 2 - 3 | 601 | 78 | |
XM_006241395.2 | XP_006241457.1 | 2 - 3 | 616 | 78 | ||
GenBank mRNA | AY030278.1 | - | 2 - 3 | 601 | 78 | |
BC072473.1 | - | 2 - 3 | 427 | 7100360 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |