Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053738.1 | NP_446190.1 | 6 - 7 | 1039 | 73 | |
XM_006241395.2 | XP_006241457.1 | 6 - 7 | 1054 | 73 | ||
GenBank mRNA | AY030278.1 | - | 6 - 7 | 1039 | 73 | |
BC072473.1 | - | 6 - 7 | 865 | 7100360 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |