Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057101.2 | NP_476442.2 | 5 - 6 | 649 | 93 | |
XM_006255907.2 | XP_006255969.1 | 5 - 6 | 723 | 93 | ||
GenBank mRNA | U56853.1 | - | 5 - 6 | 650 | 93 | |
U56854.1 | - | 4 - 5 | 711 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |