Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130742.1 | NP_570098.1 | 9 - 10 | 1387 | 92 | |
GenBank mRNA | AF241241.1 | - | 9 - 10 | 1387 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |