Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133299.2 | NP_579833.1 | 5 - 6 | 698 | 60 | |
XM_008767180.1 | XP_008765402.1 | 5 - 6 | 704 | 60 | ||
GenBank mRNA | AF021854.1 | - | 5 - 6 | 699 | 60 | |
AF099742.1 | - | 5 - 6 | 618 | 60 | ||
BC060546.1 | - | 5 - 6 | 692 | 6889831 | 60 | |
FQ209650.1 | - | 2 - 3 | 175 | 60 | ||
FQ210848.1 | - | 2 - 3 | 175 | 60 | ||
FQ218655.1 | - | 2 - 3 | 175 | 60 | ||
FQ218673.1 | - | 2 - 3 | 175 | 60 | ||
FQ218683.1 | - | 2 - 3 | 175 | 60 | ||
FQ218690.1 | - | 2 - 3 | 175 | 60 | ||
FQ219508.1 | - | 2 - 3 | 175 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |