Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172063.1 | NP_742060.1 | 3 - 4 | 189 | 64 | |
GenBank mRNA | AB017544.1 | - | 3 - 4 | 189 | 64 | |
BC081708.1 | - | 3 - 4 | 178 | 7191447 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |