Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012516.2 | NP_036648.2 | 4 - 5 | 599 | 151 | |
GenBank mRNA | BC088165.1 | - | 4 - 5 | 599 | 7324823 | 151 |
Z50051.1 | - | 2 - 3 | 262 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |