Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013074.1 | NP_037206.1 | 1 - 2 | 281 | 68 | |
XM_008766215.1 | XP_008764437.1 | 1 - 2 | 641 | 68 | ||
GenBank mRNA | AF041246.1 | - | 1 - 2 | 281 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |