Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013161.2 | NP_037293.1 | 3 - 4 | 228 | 109 | |
GenBank mRNA | D88534.1 | - | 2 - 3 | 200 | 109 | |
D88535.1 | - | 2 - 3 | 200 | 109 | ||
M58369.1 | - | 3 - 4 | 229 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |