Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133305.2 | NP_579839.1 | 2 - 3 | 478 | 91 | |
GenBank mRNA | BC086993.1 | - | 2 - 3 | 478 | 7131933 | 91 |
L07074.1 | - | 301 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |