Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133394.1 | NP_596885.1 | 4 - 5 | 740 | 73 | |
GenBank mRNA | AY040615.1 | - | 4 - 5 | 740 | 73 | |
AY886525.1 | - | 4 - 5 | 735 | 73 | ||
BC061769.1 | - | 3 - 4 | 516 | 5621307 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |