Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133567.2 | NP_598251.1 | 5 - 6 | 589 | 90 | |
GenBank mRNA | AF123047.1 | - | 5 - 6 | 516 | 90 | |
AJ007422.1 | - | 5 - 6 | 516 | 90 | ||
AJ007616.1 | - | 5 - 6 | 516 | 90 | ||
BC099775.1 | - | 5 - 6 | 589 | 7933317 | 90 | |
U51013.1 | - | 5 - 6 | 622 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |