Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134402.3 | NP_599229.1 | 4 - 5 | 479 | 148 | |
GenBank mRNA | AF031483.1 | - | 4 - 5 | 507 | 148 | |
BC063149.1 | - | 4 - 5 | 483 | 6890706 | 148 | |
FQ228189.1 | - | 4 - 5 | 455 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |