Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138870.1 | NP_620225.1 | 4 - 5 | 632 | 100 | |
XM_006228036.2 | XP_006228098.1 | 5 - 6 | 659 | 100 | ||
GenBank mRNA | AB029495.1 | - | 4 - 5 | 632 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |