Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139104.1 | NP_620804.1 | 5 - 6 | 660 | 80 | |
GenBank mRNA | AF223678.1 | - | 5 - 6 | 660 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |