Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139324.1 | NP_647540.1 | 4 - 5 | 946 | 69 | |
GenBank mRNA | AY094093.1 | - | 4 - 5 | 946 | 69 | |
BC083175.1 | - | 4 - 5 | 928 | 7105816 | 69 | |
FQ232475.1 | - | 1 - 2 | 107 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |