Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148892.3 | NP_683690.2 | 8 - 9 | 644 | 76 | |
XM_008760287.1 | XP_008758509.1 | 8 - 9 | 639 | 76 | ||
GenBank mRNA | AF523266.1 | - | 8 - 9 | 523 | 76 | |
BC062400.1 | - | 8 - 9 | 623 | 5621572 | 76 | |
FM078919.1 | - | 2 - 3 | 422 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |