Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173109.1 | NP_775132.1 | 3 - 4 | 396 | 95 | |
XM_008762473.1 | XP_008760695.1 | 3 - 4 | 401 | 95 | ||
GenBank mRNA | AJ515239.1 | - | 3 - 4 | 396 | 95 | |
AJ515241.1 | - | 2 - 3 | 216 | 95 | ||
FQ222047.1 | - | 1 - 2 | 154 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |