Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012591.1 | NP_036723.1 | 6 - 7 | 868 | 87 | |
GenBank mRNA | BC076382.1 | - | 6 - 7 | 781 | 7099391 | 87 |
M34253.1 | - | 6 - 7 | 868 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |