Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012642.4 | NP_036774.4 | 3 - 4 | 386 | 62 | |
GenBank mRNA | BC078878.1 | - | 3 - 4 | 386 | 7126969 | 62 |
J02941.1 | - | 3 - 4 | 579 | 62 | ||
S60054.1 | - | 3 - 4 | 396 | 62 | ||
X07033.1 | - | 3 - 4 | 396 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |