Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012728.1 | NP_036860.1 | 8 - 9 | 899 | 105 | |
GenBank mRNA | M97797.1 | - | 8 - 9 | 899 | 105 | |
S75952.1 | - | 8 - 9 | 893 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |