Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012860.2 | NP_036992.2 | 1 - 2 | 255 | 78 | |
GenBank mRNA | BC089770.1 | - | 1 - 2 | 255 | 7368255 | 78 |
X15734.1 | - | 1 - 2 | 162 | 78 | ||
X60822.1 | - | 1 - 2 | 341 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |