Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012865.1 | NP_036997.1 | 6 - 7 | 715 | 57 | |
GenBank mRNA | M34238.1 | - | 6 - 7 | 715 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |