Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012879.2 | NP_037011.2 | 9 - 10 | 1195 | 95 | |
XM_006232207.2 | XP_006232269.1 | 10 - 11 | 1499 | 95 | ||
GenBank mRNA | BC078875.1 | - | 9 - 10 | 1195 | 7127751 | 95 |
J03145.1 | - | 9 - 10 | 1270 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |