Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017002.2 | NP_058698.2 | 4 - 5 | 671 | 103 | |
XM_006242602.2 | XP_006242664.1 | 4 - 5 | 777 | 103 | ||
XM_006242603.2 | XP_006242665.1 | 4 - 5 | 1274 | 103 | ||
GenBank mRNA | BC089810.1 | - | 4 - 5 | 671 | 7375416 | 103 |
D13566.1 | - | 4 - 5 | 589 | 103 | ||
D83509.1 | - | 4 - 5 | 589 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |