Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017103.2 | NP_058799.2 | 2 - 3 | 194 | 111 | |
GenBank mRNA | BC081815.1 | - | 2 - 3 | 194 | 7110428 | 111 |
L46816.1 | - | 209 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |