Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017240.2 | NP_058936.1 | 23 - 24 | 3245 | 71 | |
XM_006251951.2 | XP_006252013.1 | 25 - 26 | 3395 | 71 | ||
GenBank mRNA | AB033760.1 | - | 2 - 3 | 165 | 71 | |
X15939.1 | - | 23 - 24 | 3247 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |