Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017319.1 | NP_059015.1 | 10 - 11 | 1342 | 78 | |
GenBank mRNA | BC062393.1 | - | 10 - 11 | 1379 | 5621329 | 78 |
D63378.1 | - | 10 - 11 | 1342 | 78 | ||
FQ211627.1 | - | 1441 | 78 | |||
X12355.1 | - | 10 - 11 | 1335 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |