Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019156.2 | NP_062029.2 | 2 - 3 | 263 | 75 | |
GenBank mRNA | AY318963.1 | - | 224 | 75 | ||
BC105821.1 | - | 2 - 3 | 263 | 7371104 | 75 | |
FQ209609.1 | - | 2 - 3 | 275 | 75 | ||
FQ209682.1 | - | 2 - 3 | 268 | 75 | ||
FQ210518.1 | - | 2 - 3 | 268 | 75 | ||
FQ212575.1 | - | 2 - 3 | 262 | 75 | ||
FQ218318.1 | - | 2 - 3 | 275 | 75 | ||
FQ219297.1 | - | 2 - 3 | 276 | 75 | ||
U44845.1 | - | 2 - 3 | 253 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |