Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019156.2 | NP_062029.2 | 3 - 4 | 603 | 73 | |
GenBank mRNA | BC105821.1 | - | 3 - 4 | 603 | 7371104 | 73 |
FQ209682.1 | - | 3 - 4 | 608 | 73 | ||
FQ210146.1 | - | 1 - 2 | 223 | 73 | ||
FQ210518.1 | - | 3 - 4 | 608 | 73 | ||
FQ212575.1 | - | 3 - 4 | 602 | 73 | ||
FQ218318.1 | - | 3 - 4 | 615 | 73 | ||
FQ218401.1 | - | 1 - 2 | 176 | 73 | ||
FQ219297.1 | - | 3 - 4 | 616 | 73 | ||
U44845.1 | - | 3 - 4 | 593 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |