Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019158.2 | NP_062031.1 | 4 - 5 | 726 | 71 | |
XM_006230187.2 | XP_006230249.1 | 4 - 5 | 773 | 71 | ||
GenBank mRNA | AB005547.1 | - | 4 - 5 | 725 | 71 | |
AF007775.1 | - | 4 - 5 | 705 | 71 | ||
BC081812.1 | - | 4 - 5 | 726 | 7114305 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |