Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019160.2 | NP_062033.1 | 3 - 4 | 298 | 84 | |
GenBank mRNA | AF172174.1 | - | 3 - 4 | 298 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |