Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019346.2 | NP_062219.2 | 1 - 2 | 107 | 111 | |
GenBank mRNA | FQ211495.1 | - | 1 - 2 | 140 | 111 | |
U81518.1 | - | 1 - 2 | 77 | 111 | ||
X98399.1 | - | 1 - 2 | 107 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |