Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019367.2 | NP_062240.1 | 2 - 3 | 326 | 70 | |
XM_006256017.1 | XP_006256079.1 | 2 - 3 | 641 | 70 | ||
XM_006256018.2 | XP_006256080.1 | 2 - 3 | 1463 | 70 | ||
XM_006256019.2 | XP_006256081.1 | 2 - 3 | 1196 | 70 | ||
XM_006256020.2 | XP_006256082.1 | 2 - 3 | 1051 | 70 | ||
XM_006256022.2 | XP_006256084.1 | 2 - 3 | 1010 | 70 | ||
XM_006256023.2 | XP_006256085.1 | 2 - 3 | 484 | 70 | ||
XM_008772762.1 | XP_008770984.1 | 2 - 3 | 1950 | 70 | ||
GenBank mRNA | AF061971.1 | - | 2 - 3 | 258 | 70 | |
AF067790.1 | - | 2 - 3 | 298 | 70 | ||
BC062023.1 | - | 2 - 3 | 288 | 5623515 | 70 | |
CB736242.1 | - | 2 - 3 | 381 | 70 | ||
FQ234132.1 | - | 2 - 3 | 268 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |