Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 21 - 22 | 2784 | 61 | |
GenBank mRNA | U40603.1 | - | 21 - 22 | 2784 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |