Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021700.1 | NP_067732.1 | 1 | 373 | 59 | |
GenBank mRNA | Y10619.1 | - | 1 | 373 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |