Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031608.1 | NP_113796.1 | 654 | 85 | ||
GenBank mRNA | M36418.2 | - | 457 | 85 | ||
M38060.1 | - | 457 | 85 | |||
X17184.1 | - | 654 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus 1700 Inventoried Probe spans exons |