Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181369.2 | NP_852034.1 | 4 - 5 | 2269 | 69 | |
GenBank mRNA | AY277279.1 | - | 4 - 5 | 2269 | 69 | |
FQ216289.1 | - | 4 - 5 | 2363 | 69 | ||
FQ224148.1 | - | 4 - 5 | 2355 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |