Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181371.2 | NP_852036.1 | 6 - 7 | 570 | 91 | |
XM_006236391.2 | XP_006236453.1 | 5 - 6 | 512 | 91 | ||
GenBank mRNA | FJ179400.1 | - | 6 - 7 | 536 | 91 | |
FQ211030.1 | - | 6 - 7 | 588 | 91 | ||
FQ214179.1 | - | 6 - 7 | 653 | 91 | ||
FQ220456.1 | - | 6 - 7 | 626 | 91 | ||
S83436.1 | - | 6 - 7 | 570 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |