Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181385.2 | NP_852050.1 | 1 - 2 | 797 | 86 | |
GenBank mRNA | AY283175.1 | - | 1 - 2 | 797 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |